Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193309.1 | NP_001180238.1 | 7 - 8 | 1216 | 159 | |
NM_029413.4 | NP_083689.2 | 7 - 8 | 1216 | 159 | ||
GenBank mRNA | AK017472.1 | - | 7 - 8 | 1185 | 159 | |
AK032807.1 | - | 7 - 8 | 1217 | 159 | ||
BC098483.1 | - | 7 - 8 | 1171 | 30022418 | 159 | |
BC132153.1 | - | 7 - 8 | 1070 | 40130430 | 159 | |
BC132497.1 | - | 7 - 8 | 1070 | 40130774 | 159 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |