Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145432.3 | NP_663407.2 | 3 - 4 | 560 | 119 | |
GenBank mRNA | AK012216.1 | - | 3 - 4 | 504 | 119 | |
AK076219.1 | - | 3 - 4 | 688 | 119 | ||
AK132494.1 | - | 3 - 4 | 495 | 119 | ||
AK149059.1 | - | 3 - 4 | 354 | 119 | ||
AK158323.1 | - | 3 - 4 | 690 | 119 | ||
AK169901.1 | - | 3 - 4 | 507 | 119 | ||
BC065092.1 | - | 3 - 4 | 486 | 6855410 | 119 | |
DV657360.1 | - | 3 - 4 | 560 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |