Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atad5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029856.2 | NP_001025027.1 | 9 - 10 | 3279 | 87 | |
GenBank mRNA | AY557610.1 | - | 9 - 10 | 3270 | 87 | |
BC046463.1 | - | 5 - 6 | 576 | 87 | ||
BC141180.1 | - | 8 - 9 | 2775 | 9055792 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |