Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163307.1 | NP_001156779.1 | 5 - 6 | 558 | 105 | |
NM_001163308.1 | NP_001156780.1 | 4 - 5 | 420 | 105 | ||
NM_009545.2 | NP_033571.1 | 7 - 8 | 825 | 105 | ||
GenBank mRNA | AF483502.1 | - | 3 - 4 | 266 | 105 | |
AF483503.1 | - | 3 - 4 | 266 | 105 | ||
AK142837.1 | - | 4 - 5 | 333 | 105 | ||
BC016419.1 | - | 5 - 6 | 512 | 4458285 | 105 | |
BI991588.1 | - | 4 - 5 | 486 | 105 | ||
D90085.1 | - | 7 - 8 | 673 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |