Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163307.1 | NP_001156779.1 | 7 - 8 | 717 | 79 | |
NM_001163308.1 | NP_001156780.1 | 6 - 7 | 579 | 79 | ||
NM_009545.2 | NP_033571.1 | 9 - 10 | 984 | 79 | ||
GenBank mRNA | AF483502.1 | - | 5 - 6 | 425 | 79 | |
AF483503.1 | - | 5 - 6 | 425 | 79 | ||
AK142837.1 | - | 6 - 7 | 492 | 79 | ||
BC016419.1 | - | 7 - 8 | 671 | 4458285 | 79 | |
D90085.1 | - | 9 - 10 | 832 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |