Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038619.1 | NP_001033708.1 | 4 - 5 | 802 | 74 | |
NM_172646.2 | NP_766234.1 | 4 - 5 | 806 | 74 | ||
GenBank mRNA | AK035951.1 | - | 4 - 5 | 785 | 74 | |
AK036199.1 | - | 4 - 5 | 767 | 74 | ||
AK049724.1 | - | 4 - 5 | 807 | 74 | ||
AK082698.1 | - | 4 - 5 | 746 | 74 | ||
AK147678.1 | - | 4 - 5 | 746 | 74 | ||
BC141143.1 | - | 4 - 5 | 650 | 9055755 | 74 | |
BC141144.1 | - | 4 - 5 | 650 | 9055756 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |