Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198100.2 | NP_932768.2 | 2 - 3 | 475 | 89 | |
GenBank mRNA | AK014173.1 | - | 3 - 4 | 437 | 89 | |
BC057590.1 | - | 2 - 3 | 475 | 6810986 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |