Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030731.3 | NP_109656.1 | 6 - 7 | 3068 | 103 | |
GenBank mRNA | AK029533.1 | - | 7 - 8 | 1217 | 103 | |
AK030843.1 | - | 7 - 8 | 1216 | 103 | ||
AK030862.1 | - | 6 - 7 | 3069 | 103 | ||
AK036210.1 | - | 7 - 8 | 1217 | 103 | ||
AK039280.1 | - | 7 - 8 | 1228 | 103 | ||
AK042070.1 | - | 7 - 8 | 1219 | 103 | ||
AK088922.1 | - | 6 - 7 | 1033 | 103 | ||
BC056390.1 | - | 7 - 8 | 1209 | 6854944 | 103 | |
BC059017.1 | - | 7 - 8 | 1209 | 6848690 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |