Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sun2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205345.1 | NP_001192274.1 | 10 - 11 | 1416 | 99 | |
NM_001205346.1 | NP_001192275.1 | 10 - 11 | 1410 | 99 | ||
NM_194342.3 | NP_919323.2 | 9 - 10 | 1320 | 99 | ||
GenBank mRNA | AK046318.1 | - | 9 - 10 | 1289 | 99 | |
AK156246.1 | - | 10 - 11 | 1417 | 99 | ||
AK171058.1 | - | 10 - 11 | 1379 | 99 | ||
AK172398.1 | - | 10 - 11 | 1418 | 99 | ||
AY682987.1 | - | 9 - 10 | 1176 | 99 | ||
BC098208.1 | - | 10 - 11 | 1330 | 6827666 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |