Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172842.3 | NP_766430.3 | 1 - 2 | 203 | 75 | |
GenBank mRNA | BC042765.1 | - | 1 - 2 | 203 | 4188712 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |