Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pick1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045558.1 | NP_001039023.1 | 2 - 3 | 299 | 100 | |
NM_008837.3 | NP_032863.2 | 2 - 3 | 358 | 100 | ||
GenBank mRNA | AK050224.1 | - | 2 - 3 | 270 | 100 | |
AK090155.1 | - | 291 | 100 | |||
AK162724.1 | - | 2 - 3 | 224 | 100 | ||
BC023088.1 | - | 2 - 3 | 265 | 100 | ||
BC048788.1 | - | 2 - 3 | 244 | 6488048 | 100 | |
BU057774.1 | - | 2 - 3 | 261 | 100 | ||
BU698586.1 | - | 2 - 3 | 357 | 100 | ||
CA468666.1 | - | 2 - 3 | 179 | 100 | ||
CK791908.1 | - | 2 - 3 | 299 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |