Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013672.2 | NP_038700.2 | 5 - 6 | 2144 | 75 | |
GenBank mRNA | AF022363.1 | - | 5 - 6 | 2141 | 75 | |
AF062566.1 | - | 5 - 6 | 2080 | 75 | ||
S79832.1 | - | 5 - 6 | 1193 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |