Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lonrf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028894.1 | NP_083170.1 | 6 - 7 | 1680 | 93 | |
GenBank mRNA | AK016522.1 | - | 6 - 7 | 1680 | 93 | |
AK167341.1 | - | 6 - 7 | 1738 | 94 | ||
BC120688.1 | - | 6 - 7 | 1582 | 40129611 | 93 | |
BC120690.1 | - | 6 - 7 | 1582 | 40129613 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |