Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177857.1 | NP_808525.2 | 13 - 14 | 2364 | 61 | |
GenBank mRNA | AK172232.1 | - | 9 - 10 | 1086 | 61 | |
BC046440.1 | - | 9 - 10 | 832 | 61 | ||
BC060666.1 | - | 13 - 14 | 2364 | 6841982 | 61 | |
BC066182.1 | - | 6 - 7 | 610 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |