Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030096.2 | NP_084372.2 | 11 - 12 | 1593 | 64 | |
GenBank mRNA | AK080767.1 | - | 11 - 12 | 1531 | 64 | |
AK133545.1 | - | 11 - 12 | 1564 | 64 | ||
AK141200.1 | - | 11 - 12 | 1568 | 64 | ||
AK166789.1 | - | 11 - 12 | 1519 | 64 | ||
BC029094.1 | - | 11 - 12 | 1505 | 3662249 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |