Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080453.2 | NP_536701.1 | 2 - 3 | 295 | 58 | |
NM_172797.2 | NP_766385.1 | 2 - 3 | 295 | 58 | ||
GenBank mRNA | AK051258.1 | - | 2 - 3 | 358 | 58 | |
AK151100.1 | - | 2 - 3 | 327 | 58 | ||
AY065653.1 | - | 2 - 3 | 295 | 58 | ||
AY071828.1 | - | 2 - 3 | 295 | 58 | ||
AY071829.1 | - | 2 - 3 | 295 | 58 | ||
BC137921.1 | - | 2 - 3 | 250 | 8860940 | 58 | |
BC145048.1 | - | 2 - 3 | 250 | 9053596 | 58 | |
BC145049.1 | - | 2 - 3 | 250 | 9053597 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |