Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172562.3 | NP_766150.1 | 6 - 7 | 563 | 71 | |
GenBank mRNA | AK142262.1 | - | 6 - 7 | 595 | 71 | |
AK157938.1 | - | 6 - 7 | 556 | 71 | ||
BC005610.1 | - | 3 - 4 | 290 | 71 | ||
BC025448.1 | - | 1 - 2 | 55 | 71 | ||
BC038821.1 | - | 6 - 7 | 566 | 3990023 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |