Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177503.1 | NP_001170974.1 | 11 - 12 | 1482 | 95 | |
GenBank mRNA | AK028383.1 | - | 11 - 12 | 1432 | 95 | |
BC137602.1 | - | 11 - 12 | 1265 | 8860621 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |