Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145447.2 | NP_663422.1 | 5 - 6 | 1509 | 73 | |
GenBank mRNA | AK145696.1 | - | 5 - 6 | 1466 | 73 | |
AY260574.1 | - | 5 - 6 | 1509 | 73 | ||
AY260575.1 | - | 5 - 6 | 1509 | 73 | ||
AY260576.1 | - | 5 - 6 | 1509 | 73 | ||
BC011209.1 | - | 5 - 6 | 1509 | 4190027 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |