Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009368.3 | NP_033394.2 | 2 - 3 | 1623 | 66 | |
GenBank mRNA | AK132519.1 | - | 2 - 3 | 1613 | 66 | |
AK148385.1 | - | 2 - 3 | 1487 | 66 | ||
AK162560.1 | - | 2 - 3 | 387 | 66 | ||
BC089347.1 | - | 2 - 3 | 1580 | 66 | ||
BC094591.1 | - | 3 - 4 | 1048 | 66 | ||
BC108426.1 | - | 2 - 3 | 1623 | 6757707 | 66 | |
M32745.1 | - | 2 - 3 | 1118 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |