Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK148316.1 | - | 471 | 87 | ||
AK160675.1 | - | 4 - 5 | 365 | 87 | ||
AK181546.1 | - | 3 - 4 | 87 | |||
AK183489.1 | - | 3 - 4 | 298 | 87 | ||
AK183610.1 | - | 3 - 4 | 87 | |||
AK193183.1 | - | 2 - 3 | 298 | 87 | ||
BC024483.1 | - | 2 - 3 | 103 | 87 | ||
BC027161.1 | - | 2 - 3 | 240 | 87 | ||
BC100559.1 | - | 4 - 5 | 359 | 3419074 | 87 | |
X80437.1 | - | 4 - 5 | 338 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |