Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019880.3 | NP_063933.1 | 4 - 5 | 700 | 70 | |
GenBank mRNA | AF176007.2 | - | 4 - 5 | 701 | 70 | |
AF192558.2 | - | 4 - 5 | 701 | 70 | ||
AK009623.1 | - | 4 - 5 | 583 | 70 | ||
AK154010.1 | - | 4 - 5 | 556 | 70 | ||
AK193036.1 | - | 4 - 5 | 280 | 70 | ||
AK206400.1 | - | 2 - 3 | 89 | 70 | ||
AK217430.1 | - | 4 - 5 | 281 | 70 | ||
AK219508.1 | - | 2 - 3 | 88 | 70 | ||
BC023135.1 | - | 4 - 5 | 543 | 3669288 | 70 | |
BC027274.1 | - | 4 - 5 | 549 | 3156729 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |