Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081030.2 | NP_001074499.2 | 27 - 28 | 3889 | 84 | |
NM_001170561.1 | NP_001164032.1 | 27 - 28 | 3889 | 84 | ||
GenBank mRNA | AK129485.1 | - | 27 - 28 | 3833 | 84 | |
AK147332.1 | - | 27 - 28 | 3846 | 84 | ||
BC098209.1 | - | 8 - 9 | 1232 | 84 | ||
BC157935.1 | - | 27 - 28 | 3779 | 9088115 | 84 | |
BC158013.1 | - | 27 - 28 | 3779 | 9007320 | 84 | |
BC172094.1 | - | 27 - 28 | 3779 | 9054380 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |