Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agxt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031851.1 | NP_001027021.1 | 5 - 6 | 598 | 70 | |
GenBank mRNA | AK149538.1 | - | 5 - 6 | 609 | 70 | |
BC141248.1 | - | 5 - 6 | 622 | 9055860 | 70 | |
BC145576.1 | - | 5 - 6 | 619 | 9054142 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |