Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134156.2 | NP_598917.1 | 6 - 7 | 872 | 66 | |
GenBank mRNA | AK154281.1 | - | 6 - 7 | 865 | 66 | |
BC054830.1 | - | 6 - 7 | 817 | 6308412 | 66 | |
DQ288940.1 | - | 6 - 7 | 592 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |