Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145779.1 | NP_001139251.1 | 8 - 9 | 746 | 138 | |
NM_008442.2 | NP_032468.2 | 9 - 10 | 1129 | 138 | ||
GenBank mRNA | AK016720.1 | - | 8 - 9 | 749 | 138 | |
AK037037.1 | - | 8 - 9 | 659 | 138 | ||
BC006803.1 | - | 8 - 9 | 741 | 3152883 | 138 | |
D12644.1 | - | 9 - 10 | 1130 | 138 | ||
Y15894.1 | - | 7 - 8 | 596 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |