Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgcs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291439.1 | NP_001278368.1 | 1 - 2 | 160 | 58 | |
GenBank mRNA | AK045094.1 | - | 1 - 2 | 48 | 58 | |
AK078743.1 | - | 1 - 2 | 50 | 58 | ||
AK135551.1 | - | 1 - 2 | 48 | 58 | ||
AK144384.1 | - | 1 - 2 | 48 | 58 | ||
AK146042.1 | - | 1 - 2 | 50 | 58 | ||
AK146590.1 | - | 1 - 2 | 50 | 58 | ||
BC029693.1 | - | 1 - 2 | 65 | 5375374 | 58 | |
CJ071929.1 | - | 1 - 2 | 162 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |