Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100033313.mm.1 |
NM_001080964.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033313.mm.1 |
NM_001080964.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033313.mm.1 |
NM_030220.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033313.mm.1 |
NM_030220.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033313.mm.1 |
TC1100003630.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100033319.mm.1 |
NM_001080964.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033319.mm.1 |
NM_001080964.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033319.mm.1 |
NM_030220.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033319.mm.1 |
NM_030220.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033319.mm.1 |
TC1100003630.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100033314.mm.1 |
NM_001080964.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033314.mm.1 |
NM_001080964.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033314.mm.1 |
NM_030220.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033314.mm.1 |
NM_030220.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033314.mm.1 |
TC1100003630.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100033317.mm.1 |
NM_001080964.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033317.mm.1 |
NM_001080964.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033317.mm.1 |
NM_030220.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033317.mm.1 |
NM_030220.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033317.mm.1 |
TC1100003630.mm.1 |
MTA TC |