Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165952.1 | NP_001159424.1 | 2 - 3 | 1641 | 73 | |
NM_001165953.1 | NP_001159425.1 | 2 - 3 | 1641 | 73 | ||
NM_194343.2 | NP_919324.2 | 2 - 3 | 1641 | 73 | ||
GenBank mRNA | AK076600.1 | - | 2 - 3 | 1217 | 73 | |
AK150574.1 | - | 2 - 3 | 1642 | 73 | ||
AK153571.1 | - | 2 - 3 | 1642 | 73 | ||
BC057358.1 | - | 2 - 3 | 1519 | 6835531 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |