Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR4_GL456216_random00000003.mm.1 |
NM_001033326.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_GL456216_random00000003.mm.1 |
TC4_GL456216_random00000001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR4_GL456216_random00000005.mm.1 |
NM_001033326.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_GL456216_random00000005.mm.1 |
TC4_GL456216_random00000001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR4_GL456216_random00000006.mm.1 |
NM_001033326.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_GL456216_random00000006.mm.1 |
TC4_GL456216_random00000001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR4_GL456216_random00000001.mm.1 |
NM_001033326.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_GL456216_random00000001.mm.1 |
TC4_GL456216_random00000001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR4_GL456216_random00000002.mm.1 |
NM_001033326.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_GL456216_random00000002.mm.1 |
TC4_GL456216_random00000001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR4_GL456216_random00000004.mm.1 |
NM_001033326.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_GL456216_random00000004.mm.1 |
TC4_GL456216_random00000001.mm.1 |
MTA TC |