Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100034099.mm.1 |
NM_020618.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034099.mm.1 |
NM_020618.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034099.mm.1 |
TC1100003697.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034101.mm.1 |
NM_020618.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034101.mm.1 |
NM_020618.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034101.mm.1 |
TC1100003697.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034110.mm.1 |
NM_020618.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034110.mm.1 |
NM_020618.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034110.mm.1 |
TC1100003697.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034103.mm.1 |
NM_020618.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034103.mm.1 |
NM_020618.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034103.mm.1 |
TC1100003697.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034105.mm.1 |
NM_020618.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034105.mm.1 |
NM_020618.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034105.mm.1 |
TC1100003697.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034100.mm.1 |
NM_020618.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034100.mm.1 |
NM_020618.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034100.mm.1 |
TC1100003697.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034119.mm.1 |
NM_020618.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034119.mm.1 |
NM_020618.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034119.mm.1 |
TC1100003697.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034097.mm.1 |
NM_020618.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034097.mm.1 |
NM_020618.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034097.mm.1 |
TC1100003697.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034111.mm.1 |
NM_020618.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034111.mm.1 |
NM_020618.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034111.mm.1 |
TC1100003697.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034104.mm.1 |
NM_020618.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034104.mm.1 |
NM_020618.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034104.mm.1 |
TC1100003697.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034108.mm.1 |
NM_020618.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034108.mm.1 |
NM_020618.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034108.mm.1 |
TC1100003697.mm.1 |
MTA TC |