Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146063.1 | NP_666175.1 | 6 - 7 | 1244 | 60 | |
GenBank mRNA | BC019155.1 | - | 6 - 7 | 1183 | 5065687 | 60 |
BC031593.1 | - | 6 - 7 | 1244 | 4935395 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |