Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029393.1 | NP_083669.1 | 4 - 5 | 1049 | 75 | |
GenBank mRNA | AK009986.1 | - | 3 - 4 | 394 | 75 | |
AK010165.1 | - | 4 - 5 | 1072 | 75 | ||
AK053678.1 | - | 4 - 5 | 795 | 75 | ||
BC129847.1 | - | 4 - 5 | 1049 | 40107361 | 75 | |
BC129848.1 | - | 4 - 5 | 1049 | 40107363 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |