Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zranb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285945.1 | NP_001272874.1 | 5 - 6 | 676 | 80 | |
NM_027678.3 | NP_081954.1 | 5 - 6 | 815 | 80 | ||
GenBank mRNA | AK016921.1 | - | 5 - 6 | 677 | 80 | |
AK042139.1 | - | 5 - 6 | 783 | 80 | ||
AK050039.1 | - | 1 - 2 | 96 | 80 | ||
AK083641.1 | - | 5 - 6 | 794 | 80 | ||
AK084943.1 | - | 5 - 6 | 799 | 80 | ||
BC066035.1 | - | 5 - 6 | 752 | 6837116 | 80 | |
BC117921.1 | - | 5 - 6 | 630 | 40097463 | 80 | |
BC117922.2 | - | 5 - 6 | 1822 | 40097464 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |