Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028918.2 | NP_083194.2 | 5 - 6 | 785 | 66 | |
GenBank mRNA | AK016657.1 | - | 5 - 6 | 716 | 66 | |
AK161541.1 | - | 5 - 6 | 724 | 66 | ||
AK162855.1 | - | 5 - 6 | 725 | 66 | ||
BC050805.1 | - | 5 - 6 | 809 | 6771545 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |