Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199198.2 | NP_954668.2 | 3 - 4 | 548 | 70 | |
NR_028447.1 | 3 - 4 | 548 | 70 | |||
NR_028448.1 | 3 - 4 | 548 | 70 | |||
NR_028449.1 | 3 - 4 | 548 | 70 | |||
GenBank mRNA | AK040235.1 | - | 3 - 4 | 500 | 70 | |
AK154045.1 | - | 3 - 4 | 512 | 70 | ||
BC013700.1 | - | 3 - 4 | 501 | 70 | ||
BC064018.1 | - | 3 - 4 | 475 | 3980699 | 70 | |
BU706947.1 | - | 3 - 4 | 289 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |