Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap4b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163552.1 | NP_001157024.1 | 4 - 5 | 686 | 80 | |
NM_001163553.1 | NP_001157025.1 | 3 - 4 | 899 | 80 | ||
NM_026193.2 | NP_080469.2 | 3 - 4 | 899 | 80 | ||
GenBank mRNA | AK007733.1 | - | 3 - 4 | 788 | 80 | |
AK165570.1 | - | 3 - 4 | 819 | 80 | ||
BC054092.1 | - | 4 - 5 | 697 | 30006597 | 80 | |
BC056200.2 | - | 3 - 4 | 787 | 30015292 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |