Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039684.2 | NP_001034773.2 | 5 - 6 | 540 | 78 | |
GenBank mRNA | AK030982.1 | - | 5 - 6 | 520 | 78 | |
AK138267.1 | - | 4 - 5 | 510 | 78 | ||
BC150995.1 | - | 5 - 6 | 608 | 9087908 | 78 | |
BC158122.1 | - | 5 - 6 | 700 | 9088195 | 78 | |
DQ177342.1 | - | 5 - 6 | 540 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |