Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133798.3 | NP_598559.2 | 5 - 6 | 1373 | 100 | |
GenBank mRNA | AK146901.1 | - | 5 - 6 | 1254 | 100 | |
AK147922.1 | - | 5 - 6 | 1234 | 100 | ||
AK158201.1 | - | 5 - 6 | 1264 | 100 | ||
AK162206.1 | - | 4 - 5 | 1141 | 100 | ||
AK167692.1 | - | 5 - 6 | 1374 | 100 | ||
BC018508.1 | - | 4 - 5 | 916 | 3156447 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |