Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200013989.mm.1 |
NM_001174107.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013989.mm.1 |
NM_001174107.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013989.mm.1 |
NM_177395.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013989.mm.1 |
NM_177395.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013989.mm.1 |
TC1200002029.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200013988.mm.1 |
NM_001174107.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013988.mm.1 |
NM_001174107.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013988.mm.1 |
NM_177395.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013988.mm.1 |
NM_177395.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013988.mm.1 |
TC1200002029.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200013987.mm.1 |
NM_001174107.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013987.mm.1 |
NM_001174107.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013987.mm.1 |
NM_177395.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013987.mm.1 |
NM_177395.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013987.mm.1 |
TC1200002029.mm.1 |
MTA TC |