Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010763.2 | NP_034893.1 | 3 - 4 | 1265 | 81 | |
GenBank mRNA | AK153336.1 | - | 3 - 4 | 1265 | 81 | |
BC049121.1 | - | 3 - 4 | 669 | 6813258 | 81 | |
BC068192.1 | - | 3 - 4 | 669 | 6813258 | 81 | |
U03457.1 | - | 3 - 4 | 1243 | 81 | ||
U03458.1 | - | 3 - 4 | 1243 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |