Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgfrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011197.3 | NP_035327.2 | 4 - 5 | 1240 | 110 | |
GenBank mRNA | AF152344.1 | - | 4 - 5 | 1240 | 110 | |
AK173172.1 | - | 3 - 4 | 1014 | 110 | ||
BC145713.1 | - | 4 - 5 | 1252 | 40131016 | 110 | |
BC145715.1 | - | 4 - 5 | 1252 | 40131018 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |