Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slfn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183201.4 | NP_899024.3 | 2 - 3 | 164 | 61 | |
GenBank mRNA | AK036486.1 | - | 2 - 3 | 165 | 61 | |
AK039815.1 | - | 2 - 3 | 153 | 61 | ||
AK040359.1 | - | 2 - 3 | 151 | 61 | ||
AY261804.1 | - | 1 - 2 | 126 | 61 | ||
BC117909.1 | - | 1 - 2 | 44 | 40097236 | 61 | |
BC117910.1 | - | 1 - 2 | 44 | 40097237 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |