Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016706.2 | NP_057915.2 | 4 - 5 | 1494 | 79 | |
GenBank mRNA | AA240407.1 | - | 2 - 3 | 94 | 79 | |
AF208663.1 | - | 4 - 5 | 1476 | 79 | ||
AK146366.1 | - | 4 - 5 | 1478 | 79 | ||
BC031167.1 | - | 4 - 5 | 1509 | 4983919 | 79 | |
CF105200.1 | - | 3 - 4 | 292 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |