Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011505.2 | NP_035635.1 | 14 - 15 | 1547 | 121 | |
GenBank mRNA | AF152924.1 | - | 14 - 15 | 1537 | 121 | |
AK039029.1 | - | 2 - 3 | 221 | 121 | ||
AK138499.1 | - | 2 - 3 | 220 | 121 | ||
BC032881.1 | - | 14 - 15 | 1501 | 1224878 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |