Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neurl1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081656.2 | NP_001075125.1 | 2 - 3 | 577 | 138 | |
GenBank mRNA | DQ839448.1 | - | 2 - 3 | 577 | 138 | |
GQ414760.1 | - | 2 - 3 | 577 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |