Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145473.3 | NP_663448.2 | 1 - 2 | 232 | 70 | |
GenBank mRNA | AK028916.1 | - | 1 - 2 | 97 | 70 | |
AK132493.1 | - | 1 - 2 | 130 | 70 | ||
BC016109.1 | - | 1 - 2 | 73 | 4461170 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |