Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007549.2 | NP_031575.2 | 6 - 7 | 879 | 79 | |
GenBank mRNA | AK089888.1 | - | 6 - 7 | 879 | 79 | |
AK171640.1 | - | 6 - 7 | 826 | 79 | ||
BC030668.1 | - | 6 - 7 | 729 | 1178763 | 79 | |
CT010240.1 | - | 5 - 6 | 459 | 79 | ||
M30903.1 | - | 6 - 7 | 808 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |