Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172597.2 | NP_766185.1 | 1 - 2 | 175 | 68 | |
GenBank mRNA | AK017582.1 | - | 1 - 2 | 70 | 68 | |
AK034025.1 | - | 1 - 2 | 164 | 68 | ||
AK046466.1 | - | 1 - 2 | 176 | 68 | ||
AK164111.1 | - | 1 - 2 | 184 | 68 | ||
AK173153.1 | - | 491 | 68 | |||
BC052488.1 | - | 1 - 2 | 51 | 6438726 | 68 | |
BC060101.1 | - | 1 - 2 | 177 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |