Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175413.3 | NP_780622.1 | 8 - 9 | 1179 | 87 | |
GenBank mRNA | AK039277.1 | - | 8 - 9 | 1176 | 87 | |
AK052159.1 | - | 8 - 9 | 1131 | 87 | ||
AK079125.1 | - | 8 - 9 | 1157 | 87 | ||
BC119529.2 | - | 8 - 9 | 1128 | 40056120 | 87 | |
BC119530.1 | - | 8 - 9 | 1129 | 40056121 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |